Learn More
Microsolv Technology Corporation Micro-Sample Management System. This set includes 96, 350ul Precision, Glass Inserts in a Rack with a Base and a Pre-Slit Silicone Rubber, Plug Style Sealing Mat. 1/PK.
Catalog No. 502270015
Encompass_Preferred
This Set is a convenient way to purchase all that is needed to manage your Micro-Samples or use this set in a 96 Well Plate configuration. Inserts are 1st Hydrolytic, Class A Type 33 Borosilicate Glass. Use this set with Aqueous Solutions. These 350ul inserts can easily be removed from the rack and placed in a 12x32mm, 2ml AQ™ Brand autosampler vial for use in GC or HPLC. Set includes Pre-Racked Inserts, Base & Pre-Slit Sealing Mat.
This item is not returnable. View return policy
Catalog No. 50-227-0015 Supplier Microsolv Technology Corporation Supplier No. 952963503P
This Set is a convenient way to purchase all that is needed to manage your Micro-Samples or use this set in a 96 Well Plate configuration. Inserts are 1st Hydrolytic, Class A Type 33 Borosilicate Glass. Use this set with Aqueous Solutions. These 350ul inserts can easily be removed from the rack and placed in a 12x32mm, 2ml AQ™ Brand autosampler vial for use in GC or HPLC. Set includes Pre-Racked Inserts, Base & Pre-Slit Sealing Mat.