Learn More
Microsolv Technology Corporation Micro-Sample Management System. This set includes 96, 1000ul Precision, Glass Inserts with wide conical points in a Rack with a Base and a Pre-Slit, Solvent Resistant, Polymeric Plug Style Sealing Mat. 1/PK.
Catalog No. 502270012
Encompass_Preferred
This Set is a convenient way to purchase all that is needed to manage your Micro-Samples or use this set in a 96 Well Plate configuration. 1ml Inserts are 1st Hydrolytic, Class A Type 33 Borosilicate Glass. Use with most HPLC compatible, organic solvents. These 1ml Inserts have a wide conical point for fast sample filling to minimize bubble formation and has outer dimensions of 7.72mm x 44.85mm. Set includes Pre-Racked Inserts, Base & Pre-Slit Sealing Mat.
This item is not returnable. View return policy
Catalog No. 50-227-0012 Supplier Microsolv Technology Corporation Supplier No. 952961ML3R
This Set is a convenient way to purchase all that is needed to manage your Micro-Samples or use this set in a 96 Well Plate configuration. 1ml Inserts are 1st Hydrolytic, Class A Type 33 Borosilicate Glass. Use with most HPLC compatible, organic solvents. These 1ml Inserts have a wide conical point for fast sample filling to minimize bubble formation and has outer dimensions of 7.72mm x 44.85mm. Set includes Pre-Racked Inserts, Base & Pre-Slit Sealing Mat.