Learn More
Electron Microscopy Sciences Micro Sample Tubes, 1.5ul
SDP
Catalog No. 50949245
Encompass_Preferred
Polypropylene tubes with a captive plug suitable for use in the mini centrifuge. They are steam autoclavable at 250 degree F (121 degree C).
Catalog No. 50-949-245 Supplier Electron Microscopy Sciences Supplier No. 72363
Polypropylene tubes with a captive plug suitable for use in the mini centrifuge. They are steam autoclavable at 250 degree F (121 degree C).