Learn More
Electron Microscopy Sciences Feather™ Stainless Steel Micro Scalpel 15°, Plastic Handle
SDP
Catalog No. 501899741
Encompass_Preferred
These disposable stainless steal MicroScalpels feature micro precision, ultra sharp points. They are sterilized by gamma radiation. Sure-grip aluminum handle that is color coded for easy angle identification. Comes in carbonate plastic case. Plastic handle.
Angle: 15°
Catalog No. 50-189-9741 Supplier Electron Microscopy Sciences Supplier No. 7204615
These disposable stainless steal MicroScalpels feature micro precision, ultra sharp points. They are sterilized by gamma radiation. Sure-grip aluminum handle that is color coded for easy angle identification. Comes in carbonate plastic case. Plastic handle.
Angle: 15°