Learn More
Med Vet International Micro Scalpel Blade Handle, Handle Only, J0890
Catalog No. 5000585241
Encompass_Preferred
Micro Scalpel Blades. These fine Swann Morton blades are less than half the size of standard scalpel blades. Useful for fine delicate surgery. Similar to Beaver blades. Sterile. 25/box. Autoclavable.Scalpel Handle.
Catalog No. 50-005-85241 Supplier Med Vet International Supplier No. JOR890
Micro Scalpel Blades. These fine Swann Morton blades are less than half the size of standard scalpel blades. Useful for fine delicate surgery. Similar to Beaver blades. Sterile. 25/box. Autoclavable.Scalpel Handle.