Learn More
Med Vet International Micro Scalpel Blades, #64 Micro Scalpel Blade, J0891
Catalog No. 5000584887
Encompass_Preferred
Micro Scalpel Blades. These fine Swann Morton blades are less than half the size of standard scalpel blades. Useful for fine delicate surgery. Similar to Beaver blades. Sterile. 25/box. Autoclavable.Blade. #64. Similar to standard #10 blade.
Catalog No. 50-005-84887 Supplier Med Vet International Supplier No. JOR891
Micro Scalpel Blades. These fine Swann Morton blades are less than half the size of standard scalpel blades. Useful for fine delicate surgery. Similar to Beaver blades. Sterile. 25/box. Autoclavable.Blade. #64. Similar to standard #10 blade.