Learn More
World Precision Instrument Micro Scissors, 12 cm, Diamond Coated Blades, Curved
SDP
Catalog No. 502539489
Encompass
The special diamond coated blades will increase the longevity of the cutting edge and improve the qulaity of the cut. Used to reduce wear on razor blades and industrial metal cutting tools, this coating is now being used in biomedical applications. Some studies have shown that even after double the normal usage of the cutting implement, no blade wear was detected.
Catalog No. 50-253-9489 Supplier World Precision Instrument Supplier No. 503364
The special diamond coated blades will increase the longevity of the cutting edge and improve the qulaity of the cut. Used to reduce wear on razor blades and industrial metal cutting tools, this coating is now being used in biomedical applications. Some studies have shown that even after double the normal usage of the cutting implement, no blade wear was detected.