Learn More
MSC Clarke Vantage 14 Micro Scrubber, 14.5 in, Battery, 1 ea
Catalog No. 502869010
Encompass
The Vantage 14 Micro Scrubber is a battery-operated floor scrubber designed for daily cleaning in small areas. It features a patented rotating deck for complete scrubbing in both forward and backward directions, ensuring efficient cleaning under and around obstacles. Battery-powered operation with an onboard charger and a compact design allow for maneuverability and reduced downtime.
  • Patented rotating deck enables complete scrubbing in forward and backward directions
  • Battery-powered operation with onboard charger and up to 100 minutes run time
  • Compact, maneuverable design for cleaning in limited spaces
  • Removable 3 gal solution and recovery tanks for easy filling, draining, and cleaning
  • All components, including squeegee, accessible without tools
  • Quiet operation at 64 dB A for effective daytime cleaning
Catalog No. 50-286-9010 Supplier MSC Supplier No. 35707116
Add to Cart
Add to Cart
Request a Quote
The Vantage 14 Micro Scrubber is a battery-operated floor scrubber designed for daily cleaning in small areas. It features a patented rotating deck for complete scrubbing in both forward and backward directions, ensuring efficient cleaning under and around obstacles. Battery-powered operation with an onboard charger and a compact design allow for maneuverability and reduced downtime.
  • Patented rotating deck enables complete scrubbing in forward and backward directions
  • Battery-powered operation with onboard charger and up to 100 minutes run time
  • Compact, maneuverable design for cleaning in limited spaces
  • Removable 3 gal solution and recovery tanks for easy filling, draining, and cleaning
  • All components, including squeegee, accessible without tools
  • Quiet operation at 64 dB A for effective daytime cleaning