Learn More
Fine Science Tools Moria Iris Forceps 10cm
SDP
Catalog No. NC9543845
Encompass
Style: MC31/ Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.4mm/ Alloy / Material: Inox/ Length: 10cm
Catalog No. NC9543845 Supplier Fine Science Tools Supplier No. 1137031
Style: MC31/ Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.4mm/ Alloy / Material: Inox/ Length: 10cm