Learn More
Fine Science Tools Micro Serrefines, 16mm
SDP
Catalog No. NC0451422
Encompass
Product: Micro Serrefine - Straight/16mm/ Shape: Straight/ Jaw Dimensions: 8 x 2mm/ Clamping Pressure: Light/ Gram Pressure: 85g/ Alloy: Stainless Steel/ Length: 16mm/ Serrated: Yes/ Unit: Each
Catalog No. NC0451422 Supplier Fine Science Tools Supplier No. 1805502
Product: Micro Serrefine - Straight/16mm/ Shape: Straight/ Jaw Dimensions: 8 x 2mm/ Clamping Pressure: Light/ Gram Pressure: 85g/ Alloy: Stainless Steel/ Length: 16mm/ Serrated: Yes/ Unit: Each