Learn More
Fine Science Tools Micro Serrefines, Jaw: 4 x 0.75mm, 13 mm
SDP
Catalog No. NC0426537
Encompass
Product: Micro Serrefine - Straight/13mm/ Shape: Straight/ Jaw Dimensions: 4 x 0.75mm/ Clamping Pressure: Strong/ Gram Pressure: 125g/ Alloy: Stainless Steel/ Length: 13mm/ Serrated: Yes/ Unit: Each
Catalog No. NC0426537 Supplier Fine Science Tools Supplier No. 1805504
Product: Micro Serrefine - Straight/13mm/ Shape: Straight/ Jaw Dimensions: 4 x 0.75mm/ Clamping Pressure: Strong/ Gram Pressure: 125g/ Alloy: Stainless Steel/ Length: 13mm/ Serrated: Yes/ Unit: Each