Learn More
Fine Science Tools Micro Serrefines, 15mm
SDP
Catalog No. NC9736356
Encompass
Product: Micro Serrefine - Straight/15mm/ Shape: Straight/ Jaw Dimensions: 6 x 1mm/ Clamping Pressure: Medium/ Gram Pressure: 100g/ Alloy: Stainless Steel/ Length: 15mm/ Serrated: Yes/ Unit: Each
Catalog No. NC9736356 Supplier Fine Science Tools Supplier No. 1805503
Product: Micro Serrefine - Straight/15mm/ Shape: Straight/ Jaw Dimensions: 6 x 1mm/ Clamping Pressure: Medium/ Gram Pressure: 100g/ Alloy: Stainless Steel/ Length: 15mm/ Serrated: Yes/ Unit: Each