Learn More
LUNA NANOTECH INC Micro Silica Spin Columns, 200 Columns
Supplier:  LUNA NANOTECH INC USP003200N
Encompass_Preferred
Have you ever ran out of columns that come with your favorite nucleic acid extraction kit but still had a lot of left-over buffer reagents remaining? Instead of buying a new expensive kit you can use PuroSPIN™ columns with the left-over buffers as a cost-effective alternative. Simply use PuroSPIN™ columns while following the manufacturer-specific protocols.PuroSPIN™ MICRO silica spin columns bind both DNA and RNALow 20 ?L elution volume - Maximizes concentration of purified product20 ?g nucleic acid binding capacityCompatible as cost-effective replacement columns for nucleic acid extraction kits from Qiagen, ThermoFisher, Bio-Rad, Promega and other manufacturersEach item includes both the spin column and a 2 mL sample collection tubeExtraction of DNA from bacteria, viruses, yeast, cultured cells, blood, urine, buccal swabs, animal tissue
Catalog No. 50-256-6910