Learn More
Electron Microscopy Sciences MICRO-SIZD PLSTC VIALS 30.16ML
SDP
Catalog No. 502413083
Encompass_Preferred
These one piece micro-sized hinged plastic round vials are made from Medical-Grade Polypropylene. Perfect in labs settings for dry or wet samples. They are air and moisture resistant. Have non-allergenis animal-free components and autoclavable (sterilizable) in the presence of aqueous solutions. Food packaging safe, and recyclable.
Catalog No. 50-241-3083 Supplier Electron Microscopy Sciences Supplier No. 7261744D
These one piece micro-sized hinged plastic round vials are made from Medical-Grade Polypropylene. Perfect in labs settings for dry or wet samples. They are air and moisture resistant. Have non-allergenis animal-free components and autoclavable (sterilizable) in the presence of aqueous solutions. Food packaging safe, and recyclable.