Learn More
Research Products International Corp Micro-Size Pipette Tips, Non-Sterile, 0.5 - 10ul Capacity, Bulk, 1000 per Case
SDP
Catalog No. 50212824
Encompass
These tips are designed for use on regular air displacement pipettors. Eliminates the requirement of having expensive positive displacement pipettes for critical pipetting procedures. This style is an ultra microcapillary tip that can be used for gel loading. Tip capacity is 0.5-10ul. Fits Pipetman P10 and Rainin EP-10 models.
Fits all major brands.
Catalog No. 50-212-824 Supplier Research Products International Corp Supplier No. 145104
These tips are designed for use on regular air displacement pipettors. Eliminates the requirement of having expensive positive displacement pipettes for critical pipetting procedures. This style is an ultra microcapillary tip that can be used for gel loading. Tip capacity is 0.5-10ul. Fits Pipetman P10 and Rainin EP-10 models.
Fits all major brands.