Learn More
Research Products International Corp Micro-Size Pipette Tips, Non-Sterile, 0.5 - 10 ul, Bulk, 1000 per Case
SDP
Catalog No. 50212827
Encompass
These tips are designed for use on regular air displacement pipettors. Eliminates the requirement of having expensive positive displacement pipettes for critical pipetting procedures. This style is designed for use with Eppendorf Ultramicro, Socorex, and Oxford Benchmate models. Tip capacity is 0.5-10µl. Tips are molded from high density natural polypropylene and are autoclavable.
Fits all major brands.
Catalog No. 50-212-827 Supplier Research Products International Corp Supplier No. 145204
These tips are designed for use on regular air displacement pipettors. Eliminates the requirement of having expensive positive displacement pipettes for critical pipetting procedures. This style is designed for use with Eppendorf Ultramicro, Socorex, and Oxford Benchmate models. Tip capacity is 0.5-10µl. Tips are molded from high density natural polypropylene and are autoclavable.
Fits all major brands.