Learn More
Research Products International Corp Micro-Size Pipette Tips, Sterile, 0.5 - 10 ul Capacity, 96 Tips per Rack, 960 per Case
SDP
Catalog No. 50212826
Encompass
These tips are designed for use on regular air displacement pipettors. Eliminates the requirement of having expensive positive displacement pipettes for critical pipetting procedures. This style is an ultra microcapillary tip that can be used for gel loading. Tip capacity is 0.5-10ul. Fits Pipetman P10 and Rainin EP-10 models.
Fits all major brands.
Catalog No. 50-212-826 Supplier Research Products International Corp Supplier No. 145106
Add to Cart
Add to Cart
These tips are designed for use on regular air displacement pipettors. Eliminates the requirement of having expensive positive displacement pipettes for critical pipetting procedures. This style is an ultra microcapillary tip that can be used for gel loading. Tip capacity is 0.5-10ul. Fits Pipetman P10 and Rainin EP-10 models.
Fits all major brands.