Learn More
Electron Microscopy Sciences MICRO-SIZED PLSTC VIALS 0.67ML
SDP
Supplier:  Electron Microscopy Sciences 7261741D
Encompass_Preferred
These one piece micro-sized hinged plastic round vials are made from Medical-Grade Polypropylene. Perfect in labs settings for dry or wet samples. They are air and moisture resistant. Have non-allergenis animal-free components and autoclavable (sterilizable) in the presence of aqueous solutions. Food packaging safe, and recyclable.
Catalog No. 50-241-3115