Learn More
Ibidi USA µ-Slide 2 Well, Uncoated - Uncoated Polymer Coverslip
SDP
Catalog No. NC1731059
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide 2 Well is a convenient chambered coverslip with 2 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC1731059 Supplier Ibidi USA Supplier No. 80281
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide 2 Well is a convenient chambered coverslip with 2 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box