Learn More
Electron Microscopy Sciences Micro Slide Field Finder, 75 x 25 mm
SDP
Catalog No. 50363747
Encompass_Preferred
Lovins Micros-Slide Field Finder is used to index points of interest on a microscope slide the so the fields can be relocated. A standard 75 x 25 mm glass slide has a photo etched grid pattern of 1 mm squares, subdivided into 0.1 mm intervals. Each square is marked with a letter and a number. The slide has been carefully edge ground to provide straight edges and square corners to ensure repeatable results. The pattern is imaged in vacuum deposited chrome for durability.
Size: 75 x 25 mm
Catalog No. 50-363-747 Supplier Electron Microscopy Sciences Supplier No. 7226601
Lovins Micros-Slide Field Finder is used to index points of interest on a microscope slide the so the fields can be relocated. A standard 75 x 25 mm glass slide has a photo etched grid pattern of 1 mm squares, subdivided into 0.1 mm intervals. Each square is marked with a letter and a number. The slide has been carefully edge ground to provide straight edges and square corners to ensure repeatable results. The pattern is imaged in vacuum deposited chrome for durability.
Size: 75 x 25 mm