Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abraxis Microcystins Strip Test, 20 tests, Dipstick format, Source drinking water analysis, 0-5 ppb detection range, Immunochromatographic test
SDP

Catalog No. NC1394970
Encompass

The Gold Standard Diagnostics Microcystins Strip Test is a rapid immunochromatographic assay designed for screening Microcystins and Nodularins in source drinking water. The test incorporates the QuikLyse feature for quick cell lysis, which allows for accurate measurement of total Microcystins, encompassing both dissolved and cell-bound forms. This dipstick test provides results within a range of 0-5 ppb, making it a reliable option for water quality testing. Positive results should be confirmed with additional testing methods like ELISA or HPLC.

  • Rapid immunochromatographic assay
  • Tests for Microcystins, Nodularins, and congeners
  • Designed for source drinking water
  • Features the QuikLyse technology
  • Delivers qualitative and semi-quantitative results
  • Detection range of 0-5 ppb
  • Contains 20 tests per kit

Catalog No. NC1394970 Supplier Abraxis Supplier No. 520020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Gold Standard Diagnostics Microcystins Strip Test is a rapid immunochromatographic assay designed for screening Microcystins and Nodularins in source drinking water. The test incorporates the QuikLyse feature for quick cell lysis, which allows for accurate measurement of total Microcystins, encompassing both dissolved and cell-bound forms. This dipstick test provides results within a range of 0-5 ppb, making it a reliable option for water quality testing. Positive results should be confirmed with additional testing methods like ELISA or HPLC.

  • Rapid immunochromatographic assay
  • Tests for Microcystins, Nodularins, and congeners
  • Designed for source drinking water
  • Features the QuikLyse technology
  • Delivers qualitative and semi-quantitative results
  • Detection range of 0-5 ppb
  • Contains 20 tests per kit

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.