Learn More
Ace Glass, Inc. MICRO SOXHLET UNIT C MICRO SOXHLET UNIT C
SDP
Catalog No. 50973419
Encompass_Preferred
Complete Micro Soxhlet Extractor is designed for small chemical applications. Improved design permits greater condensing capacity. Includes condenser, extractor, and flask. The condenser jacket length is approximately 139mm. The joint between the condenser and the extractor is a 24/25 standard taper joint. The 30mL capacity flask has a 14/20 standard taper joint. Hose connections are for use with 5/16¿ or 3/8" I.D. tubing. Fabricated from borosilicate glass.
Catalog No. 50-973-419 Supplier Ace Glass, Inc. Supplier No. 677610
Complete Micro Soxhlet Extractor is designed for small chemical applications. Improved design permits greater condensing capacity. Includes condenser, extractor, and flask. The condenser jacket length is approximately 139mm. The joint between the condenser and the extractor is a 24/25 standard taper joint. The 30mL capacity flask has a 14/20 standard taper joint. Hose connections are for use with 5/16¿ or 3/8" I.D. tubing. Fabricated from borosilicate glass.