Learn More
Electron Microscopy Sciences Micro Spatula, 210 mm Long
SDP
Catalog No. 5027217
Encompass_Preferred
"PTFE" coated nickel stainless steel wire with flat ends. PTFE coated nickel stainless steel wire with flat ends, 0.105" (2.7mm) dia.x8 1/4" (210mm) long, tapered to 1/8" (3mm)
Catalog No. 50-272-17 Supplier Electron Microscopy Sciences Supplier No. 72980
"PTFE" coated nickel stainless steel wire with flat ends. PTFE coated nickel stainless steel wire with flat ends, 0.105" (2.7mm) dia.x8 1/4" (210mm) long, tapered to 1/8" (3mm)