Learn More
Fine Science Tools Micro Spatula, Straight, Tip: 17.5 x 1 x 0.25mm, 11 cm
SDP
Catalog No. NC1050447
Encompass
Product: Micro Spatula - 11cm/ Tip Dimensions: 17.5 x 1 x 0.25mm Alloy: Stainless Steel 11cm/ Feature: Each
Catalog No. NC1050447 Supplier Fine Science Tools Supplier No. 1008911
Product: Micro Spatula - 11cm/ Tip Dimensions: 17.5 x 1 x 0.25mm Alloy: Stainless Steel 11cm/ Feature: Each