Learn More
Fine Science Tools Double-Ended Micro Spatula, Tip: 25 x 2 x 0.3 mm, Stainless Steel, 13 cm
SDP
Catalog No. NC9412403
Encompass
Individual Item Name: Double Sided Micro Spatula - 13cm/ Tip Shape: Straight/Angled/ Tip Dimensions: 25 x 2 x 0.3mm/ Alloy: Stainless Steel/ Length: 13cm
Catalog No. NC9412403 Supplier Fine Science Tools Supplier No. 1009112
Individual Item Name: Double Sided Micro Spatula - 13cm/ Tip Shape: Straight/Angled/ Tip Dimensions: 25 x 2 x 0.3mm/ Alloy: Stainless Steel/ Length: 13cm