Learn More
BURKLE INC Micro spatula stainless steel, V2A, length 6.69 in (170 mm)
Catalog No. 5000025733
Encompass_Preferred
The micro spatula can be used for reliably measuring, sampling or processing small amounts of powder, chemicals, pastes, creams or active ingredients. The laboratory scoop is made of stainless steel and is manufactured entirely without edges, crevices or undercuts. This excludes the possibility of impurities or cross-contamination, since no product residue or cleaning agents can be deposited.
The micro spatula is ideal for use in the pharmaceutical industry as well as food & beverage and cosmetics.
Catalog No. 50-000-25733 Supplier BURKLE INC Supplier No. 53860001
The micro spatula can be used for reliably measuring, sampling or processing small amounts of powder, chemicals, pastes, creams or active ingredients. The laboratory scoop is made of stainless steel and is manufactured entirely without edges, crevices or undercuts. This excludes the possibility of impurities or cross-contamination, since no product residue or cleaning agents can be deposited.
The micro spatula is ideal for use in the pharmaceutical industry as well as food & beverage and cosmetics.