Learn More
BURKLE INC Micro spatula, LaboPlast, PS, 0.03/0.01 oz. (1,0/0,25 ml)
Catalog No. 503081315
Encompass_Preferred
Double scoop spatula with rounded ends, content 1.0 and 0.25 ml, total length 180 mm.
The micro spatula can be used to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids.
The sturdy and convenient centre handle facilitates precise work.
Catalog No. 50-308-1315 Supplier BURKLE INC Supplier No. 53780032
Double scoop spatula with rounded ends, content 1.0 and 0.25 ml, total length 180 mm.
The micro spatula can be used to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids.
The sturdy and convenient centre handle facilitates precise work.