Learn More
BURKLE INC Micro-spatula, SteriPlast, PS, sterile, 0.03/0.01 oz. (1,0/0,25 ml)
Catalog No. NC2187721
Encompass_Preferred
Double scoop spatula with rounded ends, content 1.0 and 0.25 ml, total length 180 mm.
The micro spatula can be used to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids.
The sturdy and convenient centre handle facilitates precise work.
This item is not returnable. View return policy
Catalog No. NC2187721 Supplier BURKLE INC Supplier No. 53781032
Double scoop spatula with rounded ends, content 1.0 and 0.25 ml, total length 180 mm.
The micro spatula can be used to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids.
The sturdy and convenient centre handle facilitates precise work.