Learn More
Electron Microscopy Sciences Micro Spatula,Tappered End
SDP
Catalog No. 5027216
Encompass_Preferred
"PTFE" coated nickel stainless steel wire with flat ends. PTFE coated nickel stainless steel wire with flat ends, 0.156" (4mm) dia.x7 1/4" (184mm) long, tapered to 1/32" (0.8mm)
Catalog No. 50-272-16 Supplier Electron Microscopy Sciences Supplier No. 72982
"PTFE" coated nickel stainless steel wire with flat ends. PTFE coated nickel stainless steel wire with flat ends, 0.156" (4mm) dia.x7 1/4" (184mm) long, tapered to 1/32" (0.8mm)