Learn More
Ted Pella Inc Micro Spoon, 3/pack
SDP
Catalog No. NC1791464
Encompass
Micro Spoon, mirror stainless steel, spoon and flat spatula. Spoon 29mm L x 14.3mm W (1-1/8" L x 9/16" W). Flat 48mm L x 14.3mm W (1-7/8" L x 9/16" W). Overall length 229mm (9").
Catalog No. NC1791464 Supplier Ted Pella Inc Supplier No. 13500
Add to Cart
Add to Cart
Micro Spoon, mirror stainless steel, spoon and flat spatula. Spoon 29mm L x 14.3mm W (1-1/8" L x 9/16" W). Flat 48mm L x 14.3mm W (1-7/8" L x 9/16" W). Overall length 229mm (9").