Learn More
Electron Microscopy Sciences Micro Spoon Heyman Type A
SDP
Supplier:  Electron Microscopy Sciences 62411A
Encompass_Preferred
A Heyman type micro spatula made from nickel stainless steel with a glazed finish. The wire diameter is 0.103" (2.62 mm). Overall Length is 6 1/4" (159 mm). Flat end: 1 1/4" (31.35 mm) long and spoon end: 1/2" (12.5 mm) long x 1/8" (3 mm) wide.
Catalog No. 50-332-41