Learn More
Electron Microscopy Sciences Micro Spoon with Resin Coated End
SDP
Catalog No. 5029812
Encompass_Preferred
Made from nickel stainless steel and resin coated handle. Wire is 0.103" (2.62 mm) in diameter, overall length: 6.5" (165 mm), spoon end length: 1" (25 mm) x 1/4" (6.35 mm) wide.
Catalog No. 50-298-12 Supplier Electron Microscopy Sciences Supplier No. 6241001
Made from nickel stainless steel and resin coated handle. Wire is 0.103" (2.62 mm) in diameter, overall length: 6.5" (165 mm), spoon end length: 1" (25 mm) x 1/4" (6.35 mm) wide.