Learn More
Analytical Sales & Services Micro Static Mixing Tee for 1/16" OD Tubing
Catalog No. 5000804849
Encompass
Micro Static Mixing Tee (57540 without Frit) for 1/16" OD Tubing\n\n• 9.5µL Void Volume\n\n• .010" (0.25mm) Thru-Hole\n\n• Pressure rated to 5,000psi (345 bar)\n\n• PEEK and Kel-F (PCTFE) construction chemically resistant and biocompatible\n\n• Connect capillary tubing using our sleeves (sleeves are NOT required for 1/16" OD tubing)\n\n• Includes (3) 55132 MicroFerrules and (3) 59416 Fittings\n\n\n\nQty/Pkg: Each
Catalog No. 50-008-04849 Supplier Analytical Sales & Services Supplier No. 57540A
Add to Cart
Add to Cart
Micro Static Mixing Tee (57540 without Frit) for 1/16" OD Tubing\n\n• 9.5µL Void Volume\n\n• .010" (0.25mm) Thru-Hole\n\n• Pressure rated to 5,000psi (345 bar)\n\n• PEEK and Kel-F (PCTFE) construction chemically resistant and biocompatible\n\n• Connect capillary tubing using our sleeves (sleeves are NOT required for 1/16" OD tubing)\n\n• Includes (3) 55132 MicroFerrules and (3) 59416 Fittings\n\n\n\nQty/Pkg: Each