Learn More
Cole-Parmer Cole-Parmer Micro Stir Bar, 12.7mm L x 3mm dia 12.7mm L x 3mm dia.
Catalog No. NC0746859 Shop All Cole-Parmer Products
Encompass_Preferred
Strong alnico V magnets with one-piece coating of PTFE.
Catalog No. NC0746859 Supplier Cole-Parmer Supplier No. 0854503
Strong alnico V magnets with one-piece coating of PTFE.