Learn More
Electron Microscopy Sciences Premier Edge™ Micro Surgical Knife 15 degree
SDP
Catalog No. 50949426
Encompass_Preferred
Made from tool steel wire. Sharpened to microsurgical quality. The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Catalog No. 50-949-426 Supplier Electron Microscopy Sciences Supplier No. 7204715
Made from tool steel wire. Sharpened to microsurgical quality. The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.