Learn More
Electron Microscopy Sciences Premier Edge™ Micro Surgical Knife 30 degree
SDP
Catalog No. 50949427
Encompass_Preferred
Made from tool steel wire. Sharpened to microsurgical quality. The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Catalog No. 50-949-427 Supplier Electron Microscopy Sciences Supplier No. 7204730
Made from tool steel wire. Sharpened to microsurgical quality. The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.