Learn More
Electron Microscopy Sciences Premier Edge™ Micro Surgical Knife 45 degree
SDP
Catalog No. 50949428
Encompass_Preferred
Made from tool steel wire. Sharpened to microsurgical quality. The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Catalog No. 50-949-428 Supplier Electron Microscopy Sciences Supplier No. 7204745
Made from tool steel wire. Sharpened to microsurgical quality. The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.