Learn More
IDEX Health & Science Upchurch MICRO TEE ASMBLY .025IN OD TBG
Catalog No. NC1919730
Encompass_Preferred
Idex PEEK Microfilter Assembly, Diameter Inner: 0.006 in.
Catalog No. NC1919730 Supplier IDEX Health & Science Upchurch Supplier No. P775
Idex PEEK Microfilter Assembly, Diameter Inner: 0.006 in.