Learn More
Ted Pella Inc Micro-Tip Cryo Marking Pen, Black, Alcohol/Water Resistant, each
SDP
Catalog No. NC1764375
Encompass
Excellent for detailed writing, such as microcentrifuge and PCR tubes, labels, beakers, glass plates, X-ray and photographic films. Waterproof, high quality permanent ink micro-tip marker, and ultra-fine tip, 0.25mm.
Catalog No. NC1764375 Supplier Ted Pella Inc Supplier No. 2231511
Excellent for detailed writing, such as microcentrifuge and PCR tubes, labels, beakers, glass plates, X-ray and photographic films. Waterproof, high quality permanent ink micro-tip marker, and ultra-fine tip, 0.25mm.