Learn More
Basix™ Micro Tips, Universal Fit
GREENER_CHOICE
Catalog No. 13611108 Shop All Fisherbrand Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Refill
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
13-611-108 Non-sterile Refill
13-611-107 Sterile Rack
13-611-106 Non-sterile Rack
13-611-103 Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 13-611-108
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.
Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.
  • All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors and endotoxin and DNA
  • All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 μL
Sterility Non-sterile
Color Natural
Length (English) 1.25 in.
Length (Metric) 3.17cm
Clean Claims DNase, RNase, and DNA Free
Format Refill
Tip Style Micro Point, Universal