Learn More
Sklar Instruments Micro Adson Tissue Forceps, Sklar 4-3/4 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel. EA.
SDP
Catalog No. 501183911 Shop All Sklar Instruments Products
Encompass_Preferred
Micro Adson Tissue Forceps, Sklar. 4-3/4 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel. Sklar Micro Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are extremely narrow. This product is straight with smooth jaws, 1x2 teeth, and a length of 4-3/4 inches.
Catalog No. 50-118-3911 Supplier Sklar Instruments Supplier No. 982047
Micro Adson Tissue Forceps, Sklar. 4-3/4 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel. Sklar Micro Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are extremely narrow. This product is straight with smooth jaws, 1x2 teeth, and a length of 4-3/4 inches.