Learn More
Electron Microscopy Sciences Potter-Elvehjem Micro Tissue Grinder, 0.1 ml, 4 mm x 65 mm Mortor, 110 mm OAL
SDP
Catalog No. 501900345
Encompass_Preferred
Similar to the all glass Potter-Elvehjem style, but micro size is for extremely precise work. This unit is designed for delicate hand operation. Clearance between pestle and tube is 0.004" to 0.006" (0.1 – 015 mm). Made from Wheaton 33 low extractable borosilicate glass. Reservoir and pouring lip are incorporated into the design. Unit is autoclavable.
Size: 0.1 ml
Mortor: 4 mm x 65 mm
OAL: 110 mm
Catalog No. 50-190-0345 Supplier Electron Microscopy Sciences Supplier No. 6479401
Similar to the all glass Potter-Elvehjem style, but micro size is for extremely precise work. This unit is designed for delicate hand operation. Clearance between pestle and tube is 0.004" to 0.006" (0.1 – 015 mm). Made from Wheaton 33 low extractable borosilicate glass. Reservoir and pouring lip are incorporated into the design. Unit is autoclavable.
Size: 0.1 ml
Mortor: 4 mm x 65 mm
OAL: 110 mm