Learn More
Electron Microscopy Sciences Potter-Elvehjem Micro Tissue Grinder Pestle only, 0.1 ml, 4 mm x 65 mm Mortor, 110 mm OAL
SDP
Catalog No. 501900346
Encompass_Preferred
Replacement pestle for the Potter-Elvehjem Micro Tissue Grinderis is made of Wheaton 33 low extractable borosilicate glass.
Size: 0.1 ml
Mortor: 4 mm x 65 mm
OAL: 110 mm
Catalog No. 50-190-0346 Supplier Electron Microscopy Sciences Supplier No. 6479470
Replacement pestle for the Potter-Elvehjem Micro Tissue Grinderis is made of Wheaton 33 low extractable borosilicate glass.
Size: 0.1 ml
Mortor: 4 mm x 65 mm
OAL: 110 mm