Learn More
Cole-Parmer Master Appliance MT-51B Microtorch with Butane Fuel
Catalog No. NC0199833 Shop All Cole-Parmer Products
Encompass_Preferred
This refillable microtorch produces an adjustable wind resistant flame with temperatures up to 2500°F (1300°C). The pinpoint flame is perfect for heat shrinking and terminating as well as general heating applications. The torch features a built-in self-igniting system with gas shut-off safety lock, a extra wide base perfect for hands-free use and a durable fuel tank containing enough fuel for up to 2 hours of usage. Torch measure 6.1" H x 3" W x 1.3" D
Catalog No. NC0199833 Supplier Cole-Parmer Supplier No. 0303051
This refillable microtorch produces an adjustable wind resistant flame with temperatures up to 2500°F (1300°C). The pinpoint flame is perfect for heat shrinking and terminating as well as general heating applications. The torch features a built-in self-igniting system with gas shut-off safety lock, a extra wide base perfect for hands-free use and a durable fuel tank containing enough fuel for up to 2 hours of usage. Torch measure 6.1" H x 3" W x 1.3" D