Learn More
Optimize Technologies OPTI-LYNX™ Trap Cartridges, C4 20µL : 200-800µL/min : 2.1x5mm, 5pk
SDP
Catalog No. 502006331
Encompass_Preferred
Combining a versatile selection of packed beds and the use of a convenient quick-connect holder, Opti-Lynx provides numerous options for chromatographers for on-line or off-line sample clean-up and pre-concentration. Two styles of holders are available: The Opti-Lynx™ Micro system offers full biocompatibility in the in-line version while the direct-connect configuration threads conveniently into all 10-32 ports. Bed volumes range from 4uL to 40uL along with a wide range of standard and custom packings. Opti-Lynx traps are the ideal tool for optimizing your technique, whether you want to separate a peptide digest from its matrix for further analyses, or prepare a dilute small molecule sample for LC injection without loss of sample. These columns may be loaded and regenerated repeatedly for maximum value. Available in packages of 5 cartridges.
Catalog No. 50-200-6331 Supplier Optimize Technologies Supplier No. 1104759TG
Combining a versatile selection of packed beds and the use of a convenient quick-connect holder, Opti-Lynx provides numerous options for chromatographers for on-line or off-line sample clean-up and pre-concentration. Two styles of holders are available: The Opti-Lynx™ Micro system offers full biocompatibility in the in-line version while the direct-connect configuration threads conveniently into all 10-32 ports. Bed volumes range from 4uL to 40uL along with a wide range of standard and custom packings. Opti-Lynx traps are the ideal tool for optimizing your technique, whether you want to separate a peptide digest from its matrix for further analyses, or prepare a dilute small molecule sample for LC injection without loss of sample. These columns may be loaded and regenerated repeatedly for maximum value. Available in packages of 5 cartridges.