Learn More
Gordon Brush Mfg Co Micro Tube Brush Set 4 Inch
SDP
Catalog No. NC3336294
Encompass_Preferred
This item must be purchased in multiples of 12 per supplier requirements.
This set of seven four-inch brushes made with stainless steel wire and white nylon filaments are designed to clean very small diameter tubes including, but not limited to: airbrush and tattoo equipment, medical and lab equipment tubes, small tube, miniature tubes and pipes, narrow tubes, small cleaning brushes and other micro tubes in various diameters. Made in USA. Brush Diameters: .051", .079", .112", .146", .184", .213", .240"
Catalog No. NC3336294 Supplier Gordon Brush Mfg Co Supplier No. 710550
This item must be purchased in multiples of 12 per supplier requirements.
This set of seven four-inch brushes made with stainless steel wire and white nylon filaments are designed to clean very small diameter tubes including, but not limited to: airbrush and tattoo equipment, medical and lab equipment tubes, small tube, miniature tubes and pipes, narrow tubes, small cleaning brushes and other micro tubes in various diameters. Made in USA. Brush Diameters: .051", .079", .112", .146", .184", .213", .240"