Learn More
Research Products International Corp Screw Cap Micro-Tubes, 2.0ml Capacity, Free Standing, Graduated, 500 per case
SDP
Catalog No. 502230572
Encompass
Ideal micro-tube for high speed centrifuging, autoclaving, boiling and freezer storage. Screw cap with O ring provides superior sealing during sample processing. Tubes manufactured from transparent polypropylene for clarity. Suitable for a wide range of temperatures - from -196°C to +121°C (not for immersion in liquid nitrogen). Uniform wall thickness withstands maximum G forces over 20,000g. Versatile and durable tube developed for secure use in multiple laboratory environments.
Certified free of RNase, DNase, DNA and PCR inhibitors
Catalog No. 50-223-0572 Supplier Research Products International Corp Supplier No. 144516GR
Ideal micro-tube for high speed centrifuging, autoclaving, boiling and freezer storage. Screw cap with O ring provides superior sealing during sample processing. Tubes manufactured from transparent polypropylene for clarity. Suitable for a wide range of temperatures - from -196°C to +121°C (not for immersion in liquid nitrogen). Uniform wall thickness withstands maximum G forces over 20,000g. Versatile and durable tube developed for secure use in multiple laboratory environments.
Certified free of RNase, DNase, DNA and PCR inhibitors