Learn More
Research Products International Corp RPI Micro-Tube Ice Rack, Polystyrene, Holds 1.5ml Tubes, 120 Capacity
SDP
Catalog No. 501977811
Encompass
Ideal rack for temperature stabilized benchtop procedures or storage. Two piece design allows convenient access for filling with water or ice. High-impact polystyrene provides excellent durability for repeated use.
Catalog No. 50-197-7811 Supplier Research Products International Corp Supplier No. H18907
Ideal rack for temperature stabilized benchtop procedures or storage. Two piece design allows convenient access for filling with water or ice. High-impact polystyrene provides excellent durability for repeated use.