Learn More
Research Products International Corp Micro-Tube Rack Container with Hinged Lid, Assorted Colors, 5 per Case
SDP
Catalog No. NC1504017
Encompass
Micro-tube rack container provides a convenient method for storage of racks. Container has a clear hinged lid that protects sample tubes from airborne contaminants and frost build-up during freezer storage. Container tray is designed for easy insertion and removal from racks (Sold separately). System allows vertical storage of numerous racks for more effienct use of storage space.
Packaged five per case.
Dimensions: 9.5 x 3.5 x 2-1/8“ H
Catalog No. NC1504017 Supplier Research Products International Corp Supplier No. 164010
Micro-tube rack container provides a convenient method for storage of racks. Container has a clear hinged lid that protects sample tubes from airborne contaminants and frost build-up during freezer storage. Container tray is designed for easy insertion and removal from racks (Sold separately). System allows vertical storage of numerous racks for more effienct use of storage space.
Packaged five per case.
Dimensions: 9.5 x 3.5 x 2-1/8“ H