Learn More
Research Products International Corp Micro-Tube Rack, 80 Tube Capacity, Fluorescent Blue Color, 5 per Case
SDP
Catalog No. 50212422
Encompass
Originally designed for use with fraction collecting equipment, these racks are widely used for storing micro-centrifuge tubes. The large 80 tube capacity allows space efficient storage of samples. Racks are molded of high density polypropylene and are autoclavable.
Racks withstand temperatures from 0°C to 135°C.
Wells are individually numbered in a 16 x 5 configuration.
Catalog No. 50-212-422 Supplier Research Products International Corp Supplier No. 164102
Originally designed for use with fraction collecting equipment, these racks are widely used for storing micro-centrifuge tubes. The large 80 tube capacity allows space efficient storage of samples. Racks are molded of high density polypropylene and are autoclavable.
Racks withstand temperatures from 0°C to 135°C.
Wells are individually numbered in a 16 x 5 configuration.